4-Hydrazinobenzoesäure

Lieferant: Molekula

13488749-25G
MOLE13488749-25GEA 0 EUR
MOLE13488749-25G
4-Hydrazinobenzoesäure
4-Hydrazinobenzoic acid
Formel: C₇H₈N₂O₂
MDL: MFCD00007581
CAS-Nummer: 619-67-0
EINECS: 210-609-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR