3-Tolylboronsäure

Lieferant: Molekula

Synonyme: 3-Methylbenzolboronsäure, Dihydroxy(3-methylphenyl)boran, (3-Methylphenyl)metaborsäure, 3-Methylphenylboronsäure

13553169-1G
MOLE13553169-1GEA 0 EUR
MOLE13553169-1G
3-Tolylboronsäure
3-Tolylboronsäure
Formel: C₇H₉BO₂
Molecular Weight: 135,96 g/mol
Schmelzpunkt: 165 °C
MDL: MFCD00040198
CAS-Nummer: 17933-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR