Tetraheptylammoniumbromid

Lieferant: Molekula
Achtung

13783946-25G
MOLE13783946-25GEA 215 EUR
MOLE13783946-25G
Tetraheptylammoniumbromid
Tetraheptylammoniumbromid
Formel: C₂₈H₆₀BrN
Molecular Weight: 490,7 g/mol
Schmelzpunkt: 96 °C
MDL: MFCD00011861
CAS-Nummer: 4368-51-8
EINECS: 224-459-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR