D(+)-Cellobiose

Lieferant: Molekula

13810609-25G
MOLE13810609-25GEA 0 EUR
MOLE13810609-25G
D(+)-Cellobiose
D(+)-Cellobiose
Formel: C₁₂H₂₂O₁₁
Lagertemperatur: Raumtemperatur
MDL: MFCD00136034
CAS-Nummer: 528-50-7
EINECS: 208-436-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR