Taurin
Lieferant: Molekula
Synonyme:
2-Aminoethanesulphonic acid
13830142-1KG
MOLE13830142-1KGEA
199
EUR
MOLE13830142-1KG
Taurin
Taurin
Formel:
C₂H₇NO₃S Molecular Weight: 125.15 g/mol Schmelzpunkt: 300 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008197 CAS-Nummer: 107-35-7 EINECS: 203-483-8 Merck Index: 13,09163 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...