3',4'-Dihydroxyacetophenon

Lieferant: Molekula

Synonyme: 1-(3,4-Dihydroxyphenyl)ethan-1-on

13833699-1G
MOLE13833699-1GEA 65.6 EUR
MOLE13833699-1G
3',4'-Dihydroxyacetophenon
3',4'-Dihydroxyacetophenon
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
MDL: MFCD00016439
CAS-Nummer: 1197-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR