Chromon-2-carbonsäure
Lieferant: Molekula
Synonyme:
4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid
13834204-25G
MOLE13834204-25GEA
0
EUR
MOLE13834204-25G
Chromon-2-carbonsäure
Chromon-2-carbonsäure
Formel:
C₁₀H₆O₄ Molecular Weight: 190,15 g/mol |
MDL:
MFCD00006838 CAS-Nummer: 4940-39-0 EINECS: 225-583-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...