2-Nitroimidazol

Lieferant: Molekula
Achtung

Synonyme: 2-Nitro-1H-imidazol

13834750-1G
MOLE13834750-1GEA 146 EUR
MOLE13834750-1G
2-Nitroimidazol
2-Nitroimidazol
Formel: C₃H₃N₃O₂
Molecular Weight: 113,08 g/mol
Schmelzpunkt: 279…284 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005185
CAS-Nummer: 527-73-1
EINECS: 208-425-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,00923

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR