DL-Penicillamin

Lieferant: Molekula

Synonyme: 2-Amino-3-mercapto-3-methylbutanoic acid

13853922-25G
MOLE13853922-25GEA 886 EUR
MOLE13853922-25G
DL-Penicillamin
DL-Penicillamin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149,21 g/mol
MDL: MFCD00004856
CAS-Nummer: 52-66-4
EINECS: 200-147-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR