DL-Methionin
Lieferant: Molekula
Synonyme:
Methioninum racemicum, Methionin racemisch, H-DL-Met-OH, (±)-2-Amino-4-(methylthio)buttersäure, (±)-Methionin, (±)-2-Amino-4-(methylmercapto)buttersäure
13855549-1KG
MOLE13855549-1KGEA
155
EUR
MOLE13855549-1KG
DL-Methionin
DL-Methionin
Formel:
C₅H₁₁NO₂S Molecular Weight: 149.21 g/mol Schmelzpunkt: ∼136 °C Dichte: 1.34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063096 CAS-Nummer: 59-51-8 EINECS: 200-432-1 Merck Index: 13,06004 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...