2,4-Dimethoxy-4'-hydroxybenzophenon
Lieferant: Molekula
Synonyme:
(2,4-Dimethoxyphenyl)(4-hydroxyphenyl)methanone
13872443-5G
MOLE13872443-5GEA
0
EUR
MOLE13872443-5G
2,4-Dimethoxy-4'-hydroxybenzophenon
2,4-Dimethoxy-4'-hydroxybenzophenon
Formel:
C₁₅H₁₄O₄ Molecular Weight: 258,27 g/mol |
MDL:
MFCD00137850 CAS-Nummer: 41351-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...