2-Methylindolin
Lieferant: Molekula
|
Achtung
|
13901024-100G
MOLE13901024-100GEA
136
EUR
MOLE13901024-100G
2-Methylindolin
2-Methylindolin
Formel:
C₉H₁₁N Molecular Weight: 133,19 g/mol Siedepunkt: 228 °C (1013 hPa) Dichte: 1,0236 g/cm³ (15 °C) |
MDL:
MFCD00005706 CAS-Nummer: 6872-06-6 EINECS: 229-971-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...