Pyridoxal Hydrochlorid
Lieferant: Molekula
Synonyme:
3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl
13958515-25G
MOLE13958515-25GEA
181
EUR
MOLE13958515-25G
Pyridoxal Hydrochlorid
Pyridoxal Hydrochlorid
Formel:
C₈H₁₀ClNO₃ Molecular Weight: 203.62 g/mol |
MDL:
MFCD00012809 CAS-Nummer: 65-22-5 EINECS: 200-602-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...