7-Benzyloxyindol

Lieferant: Molekula

Synonyme: 7-(Benzyloxy)-1H-indole

14077435-1G
MOLE14077435-1GEA 71.6 EUR
MOLE14077435-1G
7-Benzyloxyindol
7-Benzyloxyindol
Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
MDL: MFCD00037974
CAS-Nummer: 20289-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR