2-Cyan-4'-methylbiphenyl

Lieferant: Molekula

Synonyme: 4'-Methyl-[1,1'-biphenyl]-2-carbonitrile

14112202-5G
MOLE14112202-5GEA 0 EUR
MOLE14112202-5G
2-Cyan-4'-methylbiphenyl
2-Cyan-4'-methylbiphenyl
Formel: C₁₄H₁₁N
Molecular Weight: 193.26 g/mol
MDL: MFCD00151805
CAS-Nummer: 114772-53-1
EINECS: 422-310-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR