3,5-Dinitrosalicylsäure
Lieferant: Molekula
|
Achtung
|
Synonyme:
2-Hydroxy-3,5-dinitrobenzoesäure, 3,5-Dinitro-2-hydroxybenzoesäure
14125911-100G
MOLE14125911-100GEA
105
EUR
MOLE14125911-100G
3,5-Dinitrosalicylsäure
3,5-Dinitrosalicylsäure
Formel:
C₇H₄N₂O₇ Molecular Weight: 228,12 g/mol Schmelzpunkt: 170…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007104 CAS-Nummer: 609-99-4 EINECS: 210-204-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...