L-Phenylalaninol

Lieferant: Molekula

14145252-5G
MOLE14145252-5GEA 0 EUR
MOLE14145252-5G
L-Phenylalaninol
L-Phenylalaninol
Formel: C₉H₁₃NO
Siedepunkt: 278 °C (1013 hPa)
Schmelzpunkt: 91…94 °C
MDL: MFCD00004732
CAS-Nummer: 3182-95-4
EINECS: 221-674-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR