Diethylacetamidomalonat

Lieferant: Molekula

Synonyme: Diethyl 2-acetamidomalonate

14164600-500G
MOLE14164600-500GEA 0 EUR
MOLE14164600-500G
Diethylacetamidomalonat
Diethylacetamidomalonat
Formel: C₉H₁₅NO₅
Molecular Weight: 217.22 g/mol
MDL: MFCD00009146
CAS-Nummer: 1068-90-2
EINECS: 213-952-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR