3,3-Dimethylglutarsäureanhydrid

Lieferant: Molekula

Synonyme: 4,4-Dimethyldihydro-2H-pyran-2,6(3H)-dione

14541420-25G
MOLE14541420-25GEA 0 EUR
MOLE14541420-25G
3,3-Dimethylglutarsäureanhydrid
3,3-Dimethylglutarsäureanhydrid
Formel: C₇H₁₀O₃
Molecular Weight: 142,15 g/mol
MDL: MFCD00006684
CAS-Nummer: 4160-82-1
EINECS: 223-998-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR