2'-Desoxycytidin Hydrochlorid

Lieferant: Molekula

Synonyme: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

14554862-1G
MOLE14554862-1GEA 71.6 EUR
MOLE14554862-1G
2'-Desoxycytidin Hydrochlorid
2'-Desoxycytidin Hydrochlorid
Formel: C₉H₁₄ClN₃O₄
Molecular Weight: 263,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012840
CAS-Nummer: 3992-42-5
EINECS: 223-639-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR