Bromphenolblau

Lieferant: Molekula

Synonyme: 3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid

14572165-25G
MOLE14572165-25GEA 154 EUR
MOLE14572165-25G
Bromphenolblau
Bromphenolblau
Formel: C₁₉H₁₀Br₄O₅S
Molecular Weight: 669.97 g/mol
Schmelzpunkt: 273
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005875
CAS-Nummer: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR