Creatinphosphat Dinatriumsalz Tetrahydrat

Lieferant: Molekula

Synonyme: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate

14577248-5G
MOLE14577248-5GEA 155 EUR
MOLE14577248-5G
Creatinphosphat Dinatriumsalz Tetrahydrat
Creatinphosphat Dinatriumsalz Tetrahydrat
Formel: C₄H₈N₃Na₂O₅P·4H₂O
Molecular Weight: 327,14 g/mol (255,07 anhy)
Lagertemperatur: Gefrierschrank
MDL: MFCD00150192
CAS-Nummer: 71519-72-7
EINECS: 213-074-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR