Dichlor-1,3,5-triazintrion Natriumsalz Dihydrat

Lieferant: Molekula
Achtung

Synonyme: NaDCC, Troclosennatrium Dihydrat, Dichlorisocyanursäure Natriumsalz Dihydrat

14579217-1KG
MOLE14579217-1KGEA 277 EUR
MOLE14579217-1KG
Dichlor-1,3,5-triazintrion Natriumsalz Dihydrat
Dichloroisocyanuric acid sodium salt dihydrate
Formel: C₃Cl₂N₃NaO₃·2H₂O
Molecular Weight: 255,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149404
CAS-Nummer: 51580-86-0
EINECS: 220-767-7
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR