D(+)-Cystin

Lieferant: Molekula

Synonyme: (S)-(+)-Cystin, (H-D-Cys-OH)2, (2S,2'S)-(+)-3,3'-Dithiobis(2-aminopropansäure)

14591553-1G
MOLE14591553-1GEA 0 EUR
MOLE14591553-1G
D(+)-Cystin
D(+)-Cystin
Formel: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
Molecular Weight: 240,3 g/mol
Schmelzpunkt: 256…261 °C
MDL: MFCD00002610
CAS-Nummer: 349-46-2
EINECS: 206-486-2
Merck Index: 13,02811

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR