3-Methylthiophen-2-carbaldehyd

Lieferant: Molekula

Synonyme: 3-Methyl-2-thiophenecarboxaldehyde, 3-Methylthiophene-2-carbaldehyde

14621318-100G
MOLE14621318-100GEA 0 EUR
MOLE14621318-100G
3-Methylthiophen-2-carbaldehyd
3-Methylthiophen-2-carbaldehyd
Formel: C₆H₆OS
Molecular Weight: 126,18 g/mol
MDL: MFCD00005430
CAS-Nummer: 5834-16-2
EINECS: 227-418-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR