L(+)-allo-Isoleucin

Lieferant: Molekula

Synonyme: (2S,3R)-(+)-allo-Isoleucin, (2S,3R)-(+)-2-Amino-3-methylpentansäure, (2S,3R)-(+)-α-Amino-β-methylvaleriansäure

14645122-250MG
MOLE14645122-250MGEA 0 EUR
MOLE14645122-250MG
L(+)-allo-Isoleucin
L(+)-allo-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Molecular Weight: 131,17 g/mol
MDL: MFCD00066446
CAS-Nummer: 1509-34-8
EINECS: 216-142-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR