3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid

Lieferant: Molekula
Achtung

Synonyme: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide

14706101-5G
MOLE14706101-5GEA 291 EUR
MOLE14706101-5G
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formel: C₁₈H₁₆N₅BrS
Molecular Weight: 414,33 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011964
CAS-Nummer: 298-93-1
EINECS: 206-069-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR