N-(2-Acetamido)-2-aminoethansulfonsäure (ACES)

Lieferant: Molekula

14774016-1KG 14774016-100G
MOLE14774016-1KGEA 0 EUR
MOLE14774016-1KG MOLE14774016-100G
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES)
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES)
Formel: H₂NCOCH₂NHCH₂CH₂SO₃H
Schmelzpunkt: 270…295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008030
CAS-Nummer: 7365-82-4
EINECS: 230-908-4
Merck Index: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR