Formamidinacetat

Lieferant: Molekula

14836567-100G
MOLE14836567-100GEA 0 EUR
MOLE14836567-100G
Formamidinacetat
Formamidinacetat
Formel: C₃H₈N₂O₂
Molecular Weight: 104.11 g/mol
MDL: MFCD00012866
CAS-Nummer: 3473-63-0
EINECS: 222-442-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR