N-Acetyl-D-methionin
Lieferant: Molekula
Synonyme:
Ac-D-Met-OH, N-Ac-D-Met-OH, (2R)-2-acetamido-4-(methylsulfanyl)butanoic acid, N-Acetyl-D-Methionine
14918715-5G
MOLE14918715-5GEA
146
EUR
MOLE14918715-5G
N-Acetyl-D-methionin
N-Acetyl-D-methionin
Formel:
C₇H₁₃NO₃S Molecular Weight: 191.2 g/mol |
MDL:
MFCD00069962 CAS-Nummer: 1509-92-8 EINECS: 216-144-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...