5-Chlor-1-indanon

Lieferant: Molekula

14962874-5G
MOLE14962874-5GEA 0 EUR
MOLE14962874-5G
5-Chlor-1-indanon
5-Chlor-1-indanon
Formel: C₉H₇ClO
Molecular Weight: 166,61 g/mol
Siedepunkt: 150 °C (0,2 mmHg)
Schmelzpunkt: 100 °C
MDL: MFCD00041456
CAS-Nummer: 42348-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR