BIS-TRIS-Propan

Lieferant: Molekula

Synonyme: 2,2'-(Propan-1,3-diyldiimino)bis[2-(hydroxymethyl)propan-1,3-diol], 1,3-Bis(tris(hydroxymethyl)methylamino)propan

14988162-500G
MOLE14988162-500GEA 0 EUR
MOLE14988162-500G
BIS-TRIS-Propan
BIS-TRIS-Propan
Formel: C₁₁H₂₆N₂O₆
MDL: MFCD00004689
CAS-Nummer: 64431-96-5
EINECS: 264-899-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR