1,4-Dimethylpiperazin
Lieferant: Molekula
Synonyme:
sym-Dimethylpiperazine
15044534-100G
MOLE15044534-100GEA
116
EUR
MOLE15044534-100G
1,4-Dimethylpiperazin
1,4-Dimethylpiperazin
Formel:
C₆H₁₄N₂ Molecular Weight: 114,19 g/mol Siedepunkt: 131 °C (1013 hPa) Schmelzpunkt: –1 °C Dichte: 0,85 g/cm³ (20 °C) |
MDL:
MFCD00006156 CAS-Nummer: 106-58-1 EINECS: 203-412-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...