DL-Valin
Lieferant: Molekula
Synonyme:
(±)-2-Amino-3-methylbutansäure, H-DL-Val-OH, (±)-Valin, (±)-2-Aminoisovaleriansäure
15070583-100G
MOLE15070583-100GEA
71.6
EUR
MOLE15070583-100G
DL-Valin
DL-Valin
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol Siedepunkt: 217 °C (1013 hPa) Schmelzpunkt: 86 °C Dichte: 1,352 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004267 CAS-Nummer: 516-06-3 EINECS: 208-220-0 Merck Index: 12,10046 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...