2'-Deoxyguanosine monohydrate

Lieferant: Molekula

Synonyme: 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one hydrate

15163303-1G
MOLE15163303-1GEA 0 EUR
MOLE15163303-1G
2'-Deoxyguanosine monohydrate
2'-Deoxyguanosine monohydrate
Formel: C₁₀H₁₃N₅O₄·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150760
CAS-Nummer: 312693-72-4
EINECS: 213-505-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR