Kanamycinsulfat
Lieferant: Molekula
|
Gefahr
|
Synonyme:
Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
15202460-5G
MOLE15202460-5GEA
71.8
EUR
MOLE15202460-5G
Kanamycinsulfat
Kanamycin sulphate
Formel:
C₁₈H₃₈N₄O₁₅S Molecular Weight: 582,6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070253 CAS-Nummer: 25389-94-0 EINECS: 246-933-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...