Ethylthiophen-2-glyoxylat

Lieferant: Molekula

Synonyme: Ethyl 2-oxo-2-(thiophen-2-yl)acetate

15242317-25G
MOLE15242317-25GEA 0 EUR
MOLE15242317-25G
Ethylthiophen-2-glyoxylat
Ethylthiophen-2-glyoxylat
Formel: C₈H₈O₃S
Molecular Weight: 184.21 g/mol
MDL: MFCD00015538
CAS-Nummer: 4075-58-5
EINECS: 223-793-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR