1,1'-Thiocarbonyldiimidazol
Lieferant: Molekula
Synonyme:
1,1'-Thiocarbonylbis(imidazole), 1,1'-Thiocarbonylbis-1H-imidazole
15276557-25G
MOLE15276557-25GEA
0
EUR
MOLE15276557-25G
1,1'-Thiocarbonyldiimidazol
1,1'-Thiocarbonyldiimidazol
Formel:
C₇H₆N₄S Molecular Weight: 178.22 g/mol Schmelzpunkt: 105…106 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005289 CAS-Nummer: 6160-65-2 EINECS: 228-183-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...