Nicotinoylchlorid Hydrochlorid

Lieferant: Molekula

15331400-25G
MOLE15331400-25GEA 0 EUR
MOLE15331400-25G
Nicotinoylchlorid Hydrochlorid
Nicotinoylchlorid Hydrochlorid
Formel: C₆H₅Cl₂NO
Molecular Weight: 178,02 g/mol
MDL: MFCD00012820
CAS-Nummer: 20260-53-1
EINECS: 243-648-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR