2',3'-Didesoxyadenosin

Lieferant: Molekula

Synonyme: ((2S,5R)-5-(6-Amino-9H-purin-9-yl)tetrahydrofuran-2-yl)methanol

15360592-25MG
MOLE15360592-25MGEA 0 EUR
MOLE15360592-25MG
2',3'-Didesoxyadenosin
2',3'-Didesoxyadenosin
Formel: C₁₀H₁₃N₅O₂
Molecular Weight: 235,24 g/mol
MDL: MFCD00010534
CAS-Nummer: 4097-22-7
EINECS: 223-853-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR