Tetrahydrothiophen-3-on

Lieferant: Molekula

Synonyme: 4,5-Dihydrothiophen-3(2H)-on

15546870-25G
MOLE15546870-25GEA 0 EUR
MOLE15546870-25G
Tetrahydrothiophen-3-on
Tetrahydrothiophen-3-on
Formel: C₄H₆OS
Molecular Weight: 102,16 g/mol
MDL: MFCD00005412
CAS-Nummer: 1003-04-9
EINECS: 213-698-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR