8-Bromadenosin

Lieferant: Molekula

Synonyme: 8-Bromoadenine-9-β-D-ribofuranoside, 6-Amino-8-bromopurine riboside, (2R,3R,4S,5R)-2-(6-Amino-8-bromo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

15591729-1G
MOLE15591729-1GEA 127 EUR
MOLE15591729-1G
8-Bromadenosin
8-Bromadenosin
Formel: C₁₀H₁₂BrN₅O₄
Molecular Weight: 346,15 g/mol
MDL: MFCD00005733
CAS-Nummer: 2946-39-6
EINECS: 220-959-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR