N-Fmoc-L-leucin

Lieferant: Molekula

15603593-25G
MOLE15603593-25GEA 71.6 EUR
MOLE15603593-25G
N-Fmoc-L-leucin
N-Fmoc-L-leucin
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353.4 g/mol
MDL: MFCD00037133
CAS-Nummer: 35661-60-0
EINECS: 252-662-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR