L(+)-Asparagin
Lieferant: Molekula
Synonyme:
(S)-(+)-Asparagin, L(+)-2-Amino-1,4-butandisäure-4-amid, (S)-(+)-2-Amino-1,4-butandisäure-4-amid
15650008-100G
MOLE15650008-100GEA
53.4
EUR
MOLE15650008-100G
L(+)-Asparagin
L(+)-Asparagin
Formel:
H₂NCOCH₂CH(NH₂)CO₂H Molecular Weight: 132,12 g/mol Siedepunkt: 343 °C (1013 hPa) Schmelzpunkt: 234…235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064401 CAS-Nummer: 70-47-3 EINECS: 200-735-9 Merck Index: 12,00872 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...