α-Cyclodextrin

Lieferant: Molekula

15673368-25G
MOLE15673368-25GEA 110 EUR
MOLE15673368-25G
α-Cyclodextrin
α-Cyclodextrin
Formel: C₃₆H₆₀O₃₀
Molecular Weight: 972,84 g/mol
MDL: MFCD00078207
CAS-Nummer: 10016-20-3
EINECS: 233-007-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR