6-Nitroindol

Lieferant: Molekula

Synonyme: 6-Nitro-1H-indole

15704199-1G
MOLE15704199-1GEA 0 EUR
MOLE15704199-1G
6-Nitroindol
6-Nitroindol
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
MDL: MFCD00051497
CAS-Nummer: 4769-96-4
EINECS: 225-311-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR