Dextran, M.W. ~ 70,000

Lieferant: Molekula

15795395-100G
MOLE15795395-100GEA 249 EUR
MOLE15795395-100G
Dextran, M.W. ~ 70,000
Dextran
Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0
EINECS: 232-677-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR