2-Aminobenzolsulfonamid

Lieferant: Molekula

Synonyme: Anilin-2-sulfonsäureamid, 2-Aminobenzolsulfonsäureamid

15813553-25G
MOLE15813553-25GEA 0 EUR
MOLE15813553-25G
2-Aminobenzolsulfonamid
2-Aminobenzolsulfonamid
Formel: C₆H₈N₂O₂S
Molecular Weight: 172,21 g/mol
MDL: MFCD00007932
CAS-Nummer: 3306-62-5
EINECS: 221-988-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR