5-(2-Fluorphenyl)-1H-tetrazol

Lieferant: Molekula

Synonyme: 5-(2-Fluorophenyl)-1H-1,2,3,4-tetraazole

15844514-1G
MOLE15844514-1GEA 0 EUR
MOLE15844514-1G
5-(2-Fluorphenyl)-1H-tetrazol
5-(2-Fluorphenyl)-1H-tetrazol
Formel: C₇H₅FN₄
MDL: MFCD00798373
CAS-Nummer: 50907-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR