3-Methoxyphenethylamin

Lieferant: Molekula

Synonyme: 2-(3-Methoxyphenyl)ethylamine

15891434-5G
MOLE15891434-5GEA 0 EUR
MOLE15891434-5G
3-Methoxyphenethylamin
3-Methoxyphenethylamin
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
MDL: MFCD00008187
CAS-Nummer: 2039-67-0
EINECS: 218-017-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR